missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maximum Recovery Diluent

Achieve maximum recovery of microorganisms during dilution with this protective, isotonic diluent.

Brand:  Thermo Scientific™ BO0348Z

 View more versions of this product

Product Code. 12776949

  • 50.35 € / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Achieve maximum recovery of microorganisms during dilution with this protective, isotonic Thermo Scientific™ Oxoid™ Maximum Recovery Diluent.

The presence of peptone in the diluent provides viable counts to ensure accurate quantitative procedures are performed. The isotonic strength of the diluent ensures recovery of organisms from various sources which may be vulnerable in distilled water or aqueous suspensions. The low peptone concentration in the diluent reduces organism multiplication in the sample for at least 45 minutes during the dilution stage1,2

Maintain the viability of organisms during dilution process with Maximum Recovery Diluent with low levels of peptone.

  • Allows accurate quantitative procedures to be performed with minimal reduction in viable count.
  • Reduces organism multiplication in the sample for at least one hour during the dilution stage.
  • Ready-to-use.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Colorless, clear
Maximum Recovery Diluent
United Kingdom
90.0g + 1.5g
10/Pk.
2 to 25°C
Sirop bottles
Agar
Ten sirop bottles with screw cap closures in a box
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number4500580 Certificate TypeCertificate of Analysis Date27/03/2025 Catalog NumberBO0348Z
Lot Number4499278 Certificate TypeCertificate of Analysis Date20/03/2025 Catalog NumberBO0348Z
Lot Number4497714 Certificate TypeCertificate of Analysis Date17/03/2025 Catalog NumberBO0348Z
Lot Number4496457 Certificate TypeCertificate of Analysis Date12/03/2025 Catalog NumberBO0348Z
Lot Number4494473 Certificate TypeCertificate of Analysis Date28/02/2025 Catalog NumberBO0348Z
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Maximum Recovery Diluent >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.