Learn More
Thermo Scientific™ Maximum Recovery Diluent
Achieve maximum recovery of microorganisms during dilution with this protective, isotonic diluent.
Brand: Thermo Scientific™ BO0348V
Description
Achieve maximum recovery of microorganisms during dilution with this protective, isotonic Thermo Scientific™ Oxoid™ Maximum Recovery Diluent.
The presence of peptone in the diluent provides viable counts to ensure accurate quantitative procedures are performed. The isotonic strength of the diluent ensures recovery of organisms from various sources which may be vulnerable in distilled water or aqueous suspensions. The low peptone concentration in the diluent reduces organism multiplication in the sample for at least 45 minutes during the dilution stage1,2
Maintain the viability of organisms during dilution process with Maximum Recovery Diluent with low levels of peptone.
- Allows accurate quantitative procedures to be performed with minimal reduction in viable count.
- Reduces organism multiplication in the sample for at least one hour during the dilution stage.
- Ready-to-use.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
Colorless, clear | |
Maximum Recovery Diluent | |
United Kingdom | |
500.0g + 1.5g | |
10/Pk. |
2 to 25°C | |
Sirop bottles | |
Agar | |
Ten sirop bottles with screw cap closures in a box |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
4484696 | Certificate of Analysis | 26/12/2024 | BO0348V |
4483020 | Certificate of Analysis | 13/12/2024 | BO0348V |
4481812 | Certificate of Analysis | 06/12/2024 | BO0348V |
4470990 | Certificate of Analysis | 26/08/2024 | BO0348V |
4467124 | Certificate of Analysis | 25/07/2024 | BO0348V |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.