missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Super Erecta™ Top Track Stationary Intermediate Unit Kits

High-density, easy access storage

Brand:  Metro™ TTA24C

 View more versions of this product

Product Code. 15977281

  • 426.00 € / Each
Estimated Shipment: 01-09-2025
to see stock.
Explore more special offers
Add to basket

Includes:
  • 2185mm (86 in.) posts
  • Hardware for connecting stationary intermediate unit to tracks on both sides
This item is not returnable. View return policy

Description

Description

Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.

  • Stationary intermediate unit helps organize Super Erecta, Super Erecta Pro™ and Super Adjustable™ Top-Track storage systems
  • 2185mm (86 in.) high posts

Order Info

Shelves are ordered separately

Specifications

Specifications

Stationary Intermediate Unit Kit
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
2185 mm
8.5 lb.
24 in.
Steel
Chrome
86 in.
Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves
3.8 kg
61 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Metro™ Super Erecta™ Top Track Stationary Intermediate Unit Kits > Chrome-plated; Shelf width: 610mm (24 in.)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.