Learn More
Merck Millex™-FG Syringe Filter Unit, Hydrophobic PTFE, 0.2 μm
Features a hydrophobic Fluoropore™ (PTFE) membrane. Millex™-FG Syringe Filter, 0.20μm, Hydrophobic PTFE provides broad chemical compatibility and is ideal for filtering samples in organic solvents or to prevent back-streaming of atmospheric moisture in venting applications.
238.00 € - 1810.00 €
Product Code | Brand | Diameter (Metric) Filter | Description | Inlet Style | Volume (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Filter | Description | Inlet Style | Volume (Metric) | Price | Quantity & Availability | ||
10094380
|
Merck
SLFG85000 |
5 cm | Luer outlet, ethylene oxide sterilized | Stepped hose barb with female Luer slip | 100 mL |
1710.00 €
Pack of 100 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||
10421551
|
Merck
SLFG05010 |
5 cm | HB/HB | Stepped hose barb with female Luer slip | 4 L |
243.00 €
Pack of 10 |
In Stock
Log in to see stock. |
||
10267770
|
Merck
SLFGN25VS |
2.5 cm | Needle outlet, ethylene oxide sterilized | Female Luer-Lok™ | 100 mL |
250.00 €
Pack of 25 |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||
10712911
|
Merck
SLFG05000 |
5 cm | HB/HB | Stepped hose barb with female Luer slip | 4 L |
1675.00 €
Pack of 100 |
Estimated Shipment: 19-11-2024
Log in to see stock. |
||
11516515
|
Merck
SLFGL25BS |
2.5 cm | Luer outlet, ethylene oxide sterilized | Female Luer-Lok™ | 100 mL |
460.00 €
Pack of 50 |
Estimated Shipment: 19-11-2024
Log in to see stock. |
||
10628261
|
Merck
SLFG65000 |
5 cm | HB-NPTM | Stepped Hose Barb with Female Luer slip interior | 4 L |
1810.00 €
Pack of 100 |
Estimated Shipment: 19-11-2024
Log in to see stock. |
||
10410011
|
Merck
SLFG65010 |
5 cm | Inlet and Outlet, 1/8 NPT | Stepped Hose Barb with Female Luer slip interior | 4 L |
238.00 €
Pack of 10 |
Estimated Shipment: 19-11-2024
Log in to see stock. |
||
10238090
|
Merck
SLFG85010 |
5 cm | Luer outlet, ethylene oxide sterilized | Stepped hose barb with female Luer slip | 100 mL |
243.00 €
Pack of 10 |
N/A | ||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
10638261
|
Merck
SLFG75010 |
5 cm | Inlet and Outlet, 1/8 NPT | 1/8 in. NPTM | 100 mL |
238.00 €
Pack of 10 |
Estimated Shipment: 19-11-2024
Log in to see stock. |
|
10687121
|
Merck
SLFG55010 |
5 cm | HB-NPTM | Stepped hose barb with female luer slip interior | 4 L |
238.00 €
Pack of 10 |
N/A | ||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
10648261
|
Merck
SLFG75000 |
5 cm | HB-NPTM | 1/8 in. NPTM | 4 L |
1810.00 €
Pack of 100 |
Estimated Shipment: 19-11-2024
Log in to see stock. |
|
Description
Protect vacuum sources from contamination and moisture damage
Millex™ syringe filter units with hydrophobic Fluoropore™ or Millipore Express™ PLUS PES membrane are ideal for in-line sterilization of gases and venting sterile containers, and filters with Fluoropore™ membrane can also be used for sterilizing or clarifying organic solutions. There are also specialized filter units to protect hemodialysis transducers from blood and moisture. The 50 mm Millex™ filter units are especially useful for vacuum line protection. All units are bi-directional.Features & Benefits:
- Specialized filter units protect hemodialysis transducers from blood and moisture
- 50 mm Millex™ units ideal for vacuum line protection
- All Millex™ Hydrophobic PTFE units are bi-directional in flow
Applications:
Sterilizing Gases, Vacuum Line Protection, Venting Sterile Containers, Sterilizing and Clarifying Organic Solutions
Specifications
Single-use | |
0.2 μm |