Learn More
MP Biomedicals™ Histopaque™ 1077 Solution
For in vitro isolation of lymphocytes. GE Healthcare™ Histopaque™ 1077 Solution is used in areas such as histocompatability testing, in vitro cell-mediated immunity assays, and other procedures requiring pure lymphocyte preparations.
Brand: MP Biomedicals™ 0219083780
138.07 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Solution of polysaccharide (Ficoll-400) and a radiopaque contrast medium (sodium diatrizoate)
- Easy one-step, rapid, reproducible method for the preparation of viable lymphocytes in high yield

Specifications
100 mL | |
For in-vitro isolation of lymphocytes | |
1.077 g/mL |
4°C | |
Centrifugation Media | |
Sterile |
Safety and Handling
- Histopaque
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H350-May cause cancer.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : hypaque, ficoll 400
Your input is important to us. Please complete this form to provide feedback related to the content on this product.