Learn More
MP Biomedicals™ Human Serum: Normal Serum
Hepatitis Associated Antigen (HBsAG)–free, HIV–free
Brand: MP Biomedicals™ 092930149
317.18 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description

Specifications
U.S. regulations | |
-20°C | |
Liquid | |
100 mL | |
0.1 μm filtration | |
Media component for cell free virus, liposome binding assay, preparation of sperm | |
No | |
7 to 8 |
Golden Brown | |
Human | |
Human Serum | |
Sterile | |
1EU/mL | |
≤26mg/dL | |
263 to 323mOsm/kg | |
5.5 to 7.5 g/dL |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.