missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzylmethylamine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
250 mL
1000 mL
Description
N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.
Solubility
Soluble in water (65 g/L at 20°C).
Notes
Air Sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, carbon dioxide.
N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.
Solubility
Soluble in water (65 g/L at 20°C).
Notes
Air Sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, carbon dioxide.
Specifications
Specifications
Melting Point | -55°C |
Density | 0.94 |
pH | 10 |
Boiling Point | 184°C to 188°C |
Flash Point | 77°C (170°F) |
Odor | Amine-like |
Linear Formula | C6H5CH2NHCH3 |
Refractive Index | 1.523 |
Quantity | 100 mL |
UN Number | UN2922 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'