Learn More
Thermo Scientific™ Nalgene™ Black Ink Lab Markers
Mark on glass, plastic and metal with these black ink lab markers that have caps with a convenient packet clip.
Brand: Thermo Scientific™ 6310-0010
48.52 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Mark on glass, plastic and metal with Thermo Scientific™ Nalgene™ Black Ink Lab Markers, recommended for use with Nalgene™ Polyolefin paper, notebooks and labels. These markers have caps with a convenient packet clip. Choose from fine point or broad line options.
Features:
- Felt-tip marking pens with pocket clips and black ink
- Choice between fine-line pen or broad-line marker
- Writes on glass, plastic or metal
Specifications
Fine | |
Lab Pen, Fine Line | |
4 |
Black | |
12 |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
7448043010 | Certificate of Compliance | 31/03/2025 | 6310-0010 |