missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Sharpie™ Fine Point Permanent Marker
All-purpose markers with fine (1mm) tips. Cole-Parmer™ Sharpie™ Fine Point Permanent Marker feature quick-drying, water-resistant ink.
Brand: Cole-Parmer™ 09968-00
65.86 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
- Fine (1mm) tip
- Quick drying
- Water-resistant ink
- Nontoxic
- Conform to ASTM D-4236
Specifications
Permanent Marker | |
Black | |
12 Pk. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 15
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ Sharpie™ Fine Point Permanent Marker > Color: Black
Spot an opportunity for improvement?Share a Content Correction