missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 96 Well Edge Plate, TC Surface, No Lid, Pack of 10
GREENER_CHOICE

Color:Clear; Sterility:Sterile; Lid:Without Lid

Brand:  Thermo Scientific™ 167554

383.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15553115

  • 405.00 â‚¬ / Pack of 160
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate. When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste.

Features
  • The Edge 2.0 Plate provides an efficient and economical solution for preventing evaporation and in turn the risks associated with edge effect.
  • The surrounding built-in moat is divided into 4 sectional reservoirs, each can be filled with 1.7 to 2.0mL sterile water, buffer or media.
  • Replenish fluid in the reservoirs every 3 days in the humidified incubator to ensure the effectiveness of the evaporation barrier.
  • Nunclon™ Delta cell culture surface treatment of the wells promotes cell attachment and growth.
  • The non-treated surface is available for suspension cell culture or general solution-based assays
  • Use the Edge 2.0 Plate in conjunction with the Thermo Scientific™ Varioskan™ LUX multimode microplate reader. The filled moat of the plate functions as passive humidity control, allowing for simultaneous incubation and signal detection during long-term applications with living cells inside the plate reader.
Warranty: 90 days
TRUSTED_SUSTAINABILITY
Specifications

Specifications

All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Without Lid
8 x 12
Nunc
Microplate
Polystyrene
96
Nunclon Delta
Less waste, fewer resources used
10
Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
0.35 cm2/well
10 Pk., 160 Cases
Clear
Low Flange
400 ÎĽL
Sterile
Flat Bottom (F-well)
160
160 Plates
Product Suggestions

Product Suggestions

Videos

Videos

  • Thermo Scientific Nunc - Edge 2.0 plate
    Minimise plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific Nunc Edge 2.0 plate in place of your standard 96 well plate.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Thermo Scientific Nunc Edge 2.0 plate

    • Thermo Scientific Nunc - Edge 2.0 plate
      Minimise plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific Nunc Edge 2.0 plate in place of your standard 96 well plate.
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
       
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected

        Thermo Scientific Nunc Edge 2.0 plate

      SDS
      Documents Certificates

      Certificates

      A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

      2 results found
      Lot Number Certificate Type Date Catalog Number
      Lot Number185792 Certificate TypeCertificate of Compliance Date26/04/2025 Catalog Number167554
      Lot Number171763 Certificate TypeCertificate of Compliance Date02/12/2024 Catalog Number167554
      Special Offers

      Special Offers

      Product Content Correction

      Your input is important to us. Please complete this form to provide feedback related to the content on this product.

      Product Title
      Thermo Scientific™ 96 Well Edge Plate, TC Surface, No Lid, Pack of 10 > Treated, 400 uL, 160

      By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

      Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.