Learn More
Thermo Scientific™ Nunc™ Square BioAssay Dishes

Screen large numbers of colonies for cloning experiments, culturing bacteria, fungi and agar diffusion assays using Thermo Scientific™ Nunc™ Squre BioAssay dishes.
Brand: Thermo Scientific™ 166508
197.91 EUR valid until 2025-12-31
Use promo code "24345" to get your promotional price.
Description
Available in cell culture-treated and non-treated surfaces, the versatile Thermo Scientific™ Nunc™ Square BioAssay dishes are useful for screening large numbers of colonies, culturing bacteria and fungi, or providing a contained environment for other culture vessels.
- Use as a moisture chamber for incubation on Nunc MicroWell™ Plates
- Lined with filter paper it is ideal as an incubation chamber for in situ hybridization
- Optically clear and compatible with automated robotic instrumentation
Untreated Dishes
- Ideal for screening libraries or isolating clones
- Useful for screening large numbers of colonies in cloning experiments
- Can be used for culturing of bacteria and fungi as well as a moisture chamber for incubation of Nunc MicroWell Plates
- Suitable in agar diffusion assay
- Lined with wet filter paper, it is also useful as an incubation chamber for in situ hybridization
- Large growth area
- Designed for genomic screen and colony picking

Specifications
Flat | |
Clear | |
Square | |
Nunclon Delta | |
135 mL | |
4 | |
25 in. |
135 mL | |
500 sq. cm | |
Sterile | |
Multi-Well Plate | |
16 | |
25 mm |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
170717 | Certificate of Compliance | 03/12/2024 | 166508 |
173402 | Certificate of Compliance | 03/12/2024 | 166508 |
171738 | Certificate of Compliance | 03/12/2024 | 166508 |
181493 | Certificate of Compliance | 03/12/2024 | 166508 |
181494 | Certificate of Compliance | 03/12/2024 | 166508 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.