Learn More
Cytiva O-ring
Use in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules. Cytiva Life Sciences™ O-rings are made of nitrile rubber.
Brand:  Cytiva 19-7595-01
 View more versions of this product
Product Code. 11570854
  • 27.00 € / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Can be used in aqueous solutions
  • Nearly all organic solvents
  • The following few exceptions: chlorinated hydrocarbons, acetone and otherketones, aliphatic esters and phenol. Solutions containingmore than 10% NaOH, 10% HCl, other strong mineral acidsor 5% acetic acid should not be used
  • Made of Nitrile Rubber (NBR)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nitrile Rubber
5/Pk.
O-Ring
27.1 x 1.60 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva O-ring > Size: 27.1 x 1.60mm; Material: Nitrile Rubber; Quantity: 5/Pk.