Learn More
Cole-Parmer™ Lab Optic Cleaning Kit for Instruments Lenses
Keep lenses dust and particle free with this cleaning kit. Optic Cleaning Kit for Instruments Lenses is ideal for microscopes, magnifiers and lenses. The cleaning solution effectively removes oils and residues from the optic surface.
Brand: Cole-Parmer™ 46301-10
71.55 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
- Includes: StaticWisk™ anti-static brush, SpeckGRABBER™ probe, Tiger Cloth™ anti-static microfiber cleaning cloth and a 1 oz. bottle of Precision Cleaning Solution™.
Specifications
Optics Cleaning Kit | |
Kinetronics |
Safety and Handling
- isopropyl alcohol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use CO2, dry chemical, alcohol foam or waterspray to extinguish.
Supplemental information
- MIXTURE LIST-Contains : isoprpyl alcohol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.