missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brain Heart Infusion Agar (Dehydrated)

Cultivate fastidious organisms with this highly nutritious medium.

Brand:  Thermo Scientific™ CM1136B

Product Code. 11934102

  • 170.00 € / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Cultivate fastidious organisms with highly nutritious Thermo Scientific™ Oxoid™ Brain Heart Infusion Agar (Dehydrated). Brain Heart Infusion Agar is recommended for the cultivation of streptococci, Neisseria and other fastidious organisms.

Cultivate a wide variety of organisms with Brain Heart Infusion Agar.

  • Recommended for the isolation if Neisseria gonorrhoeae1.
  • With addition of blood and antibiotics the medium is suitable for the isolation of the tissue phase of Histoplasma capsulatum and other pathogenic fungi including Coccidiodes immitis2, 3.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Industrial Reference: AFNOR, BSI, FDA, IDF, ISO, USDA.
Powder
500 g
Brain Heart Infusion Agar
Agar
For 10.6L medium
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6105300 Certificate TypeCertificate of Analysis Date09/05/2024 Catalog NumberCM1136B
Lot Number6105299 Certificate TypeCertificate of Analysis Date27/04/2024 Catalog NumberCM1136B
Lot Number3780664 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog NumberCM1136B
Lot Number3716895 Certificate TypeCertificate of Analysis Date08/09/2023 Catalog NumberCM1136B
Lot Number3687148 Certificate TypeCertificate of Analysis Date06/07/2023 Catalog NumberCM1136B
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Brain Heart Infusion Agar (Dehydrated) > 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.