Learn More
R&L Enterprises™ Rectangular Retort Stand Bases
An excellent option for supporting retort stands in the laboratory. R&L Enterprises™ Rectangular Retort Stand Bases are manufactured from white acrylic finished steel.
Brand:  R&L Enterprises™ 3107/31
 View more versions of this product
Product Code. 11705386
  • 38.06 € / Each
Estimated Shipment: 02-05-2025
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Without rod
  • Fitted with rubber feet
  • Hole threaded M10 at the center of one short side
Specifications
Specifications
Stainless Steel
160 mm
Tripod Plate
250 mm
With Rubber Feet
White
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
R&L Enterprises™ Rectangular Retort Stand Bases > Base Length: 250mm; Base Width: 160mm