Learn More
Thermo Scientific™ Buffered Peptone Water (Dehydrated)
Pre-enrich food samples prior to the isolation of Salmonella species while allowing the resuscitation of sub-lethally injured cells with this medium.
Brand: Thermo Scientific™ CM0509T
Description
Pre-enrich food samples prior to the isolation of Salmonella species using Thermo Scientific™ Oxoid™ Buffered Peptone Water (Dehydrated). Buffered Peptone Water is a non-selective pre-enrichment medium. It also provides conditions for resuscitation of cells that have been injured by processes of food preservation.
Low numbers and/or sub-lethally injured Salmonella spp. cells may be present in a test food sample. Pre-enrichment in non-selective Buffered Peptone Water allows the cells time to repair and multiply before being introduced into a selective culture, improving the chances of recovering Salmonella species from the sample.
- Supports high growth and recovery rate: rich in nutrients, the medium produces high resuscitation rates and supports intense growth.
- Helps recovery of sub-lethally damaged salmonellae: the phosphate buffer system of the medium prevents bacterial damage due to changes in pH of the medium and helps the recovery of sub-lethally damaged salmonellae that may be sensitive to low pH.
It was noted by Edel and Kampelmacher1 that sub-lethal injury to salmonellae may occur in many food processes. In a survey involving isolation of salmonellae from meat that had been artificially contaminated with sub-lethally injured organisms, pre-enrichment in buffered peptone water at 37°C for 18 hours before selection in Brilliant Green Tetrathionate Bile Broth showed superior results compared with a direct selection method.
Sadovski2 reported that, in experiments involving isolation of salmonellae from frozen vegetables, the rapid drop in pH when using lactose broth3 as a pre-enrichment medium was detrimental to the recovery of salmonellae. This was due to the enhanced sensitivity to low pH of freeze-injured salmonellae which may contaminate frozen vegetables. Pre-enrichment with Buffered Peptone Water maintained a high pH over a period of 24 hours of incubation. The vegetable tissue has a low buffering capacity and the medium overcame this problem.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

Specifications
Buffered Peptone Water | |
Pre-enrichment Medium | |
For 250.0L medium |
Powder | |
5 kg |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6248481 | Certificate of Analysis | 04/04/2025 | CM0509T |
6224125 | Certificate of Analysis | 25/02/2025 | CM0509T |
6224123 | Certificate of Analysis | 25/02/2025 | CM0509T |
6230077 | Certificate of Analysis | 25/02/2025 | CM0509T |
6216180 | Certificate of Analysis | 22/01/2025 | CM0509T |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Laboratory Use Only