Learn More
Gibco™ PBS, pH 7.4
Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
Brand: Gibco™ 10010001
Description
Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation.
We offer a variety of Gibco™ PBS formulations for a range of cell culture applications.
This PBS is manufactured as follows:
Without
• Calcium
• Magnesium
• Phenol Red
The complete formulation is available.

Specifications
No Sodium Pyruvate | |
15°C to 30°C | |
Gibco™ | |
10 L | |
Mammalian Cell Culture | |
7.3 to 7.5 | |
Ambient | |
Liquid |
Animal Origin-Free | |
1 X | |
Phosphate Buffered Saline (PBS) | |
Phosphate Buffered Saline | |
280 - 315 mOsm/kg | |
No Phenol Red | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2572130 | Certificate of Analysis | 21/02/2025 | 10010002,10010001,10010023,10010031 |
2494223 | Certificate of Analysis | 21/02/2025 | 10010002,10010001,10010023,10010031 |
2494226 | Certificate of Analysis | 21/02/2025 | 10010002,10010001,10010023,10010031 |
2562446 | Certificate of Analysis | 21/02/2025 | 10010002,10010001,10010023,10010031 |
2436334 | Certificate of Analysis | 21/02/2025 | 10010002,10010001,10010023,10010031 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.