Learn More
MP Biomedicals™ Phosphate Buffered Saline Tablets (PBS)
Phosphate Buffered Saline Tablets (PBS) are a balanced salt mixture for easy PBS buffer preparation for a variety of applications, including reagent preparation, diluting cells for flow cytometry, and cell culture reagent.
Brand:  MP Biomedicals™ 092810307
429.40 EUR valid until 2025-12-31
Use promo code "24341" to get your promotional price.
 View more versions of this product
Product Code. 11455015
  • 452.00 € / 500 tablets
Explore more special offers
Description
Description
  • No need for weighing and mixing chemicals
  • Simply dissolve PBS tablets in water
  • One tablet dissolved in 100mL water yields buffer containing 0.01M phosphate, 0.0027M KCl, and 0.137M NaCl, pH 7.4 at 25°C
  • Research use only
TRUSTED_SUSTAINABILITY
Specifications
Specifications
No Calcium, No Magnesium
500 Tablets
MFCD00011360
[Cl-].[K+]
74.55
Phosphate Buffered Saline (PBS)
ClK
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
Tablet
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals™ Phosphate Buffered Saline Tablets (PBS) > Phosphate Buffered Saline Tablets (PBS), 500 Tablets