missing translation for 'onlineSavingsMsg'
Learn More

Pen-Strep (10,000 IU/mL, 10 mg/mL), MP Biomedicals™

Pen-Strep (10,000 IU/mL, 10 mg/mL) is a double antibiotic solution containing 10,000 units/mL of penicillin and 10,000 μg/mL of streptomycin.

Brand:  MP Biomedicals 091670249

36.49 EUR valid until 2025-05-31
Use promo code "24127" to get your promotional price.


 View more versions of this product

Product Code. 11490425

  • 38.70 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • The antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria.
  • This solution contains 10,000 units/mL of penicillin and 10,000 μg/mL of streptomycin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Penicillin-Streptomycin
Penicillin/Streptomycin
100 mL
200X Pen-Strep
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pen-Strep (10,000 IU/mL, 10 mg/mL), MP Biomedicals™ > Quantity: 100 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.