missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Nickel Coated Plates

Thermo Scientific™ Pierce Nickel Coated Plates provide a simple format to bind His-tagged fusion proteins for ELISA and other plate-based assays involving protein interactions of expressed recombinant proteins.

Brand:  Thermo Scientific™ 15242

 View more versions of this product

Product Code. 10033983

  • 344.00 € / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

These Ni(2+) chelate-coated plates are ideal for analyzing polyhistidine-tagged fusion proteins by ELISA-based methods. Proteins that contain a succession of several histidine residues at the amino or carboxyl terminus have a strong binding affinity for metal. Bacterial lysates containing polyhistidine-tagged fusion proteins can be added directly to the plates without the need for blocking. The clear, white or black plates can be used with colorimetric, chemiluminescent or fluorescent detection methods, respectively.

Thermo Scientific Pierce Nickel Coated White 96-Well Plates provide a simple format to bind His-tagged fusion proteins for ELISA and other plate-based assays involving protein interactions of expressed recombinant proteins. These white plates can be used with chemiluminescent detection methods.

Features of Pierce Nickel Coated Plates include:
• Ni2+ activated surface enables metal-chelate binding of His-tagged proteins
• Detergents used to lyse cells don't inhibit binding to activated plates as they do with plain polystyrene
• Better binding for sensitive assays compared to other commercially available nickel-activated plates
• Detection limit: 1 ng of polyhistidine fusion protein
• Binding capacity: approx. 9 pmol His-tagged protein (27 kDa) per well
• Activation level: 200 μL

These Ni(2+) chelate-coated plates are ideal for analyzing polyhistidine-tagged fusion proteins by ELISA-based methods. Proteins that contain a succession of several histidine residues at the amino or carboxyl terminus have a strong binding affinity for metal. Bacterial lysates containing polyhistidine-tagged fusion proteins can be added directly to the plates without the need for blocking.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard Capacity, Affinity
Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C.
5 Plates
White
Polystyrene
96
Blocker™ BSA Buffer
Standard
Chemiluminescence
Pierce
Microplate
200 μL
Divalent Nickel, Metal Chelate
5 plates
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZE394905 Certificate TypeCertificate of Analysis Date15/06/2024 Catalog Number15242
Lot NumberYJ380274 Certificate TypeCertificate of Analysis Date07/12/2023 Catalog Number15242
Lot NumberXJ360044 Certificate TypeCertificate of Analysis Date26/10/2022 Catalog Number15242
Lot NumberXA340178 Certificate TypeCertificate of Analysis Date20/01/2022 Catalog Number15242
Lot NumberWG322320 Certificate TypeCertificate of Analysis Date22/07/2021 Catalog Number15242
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Nickel Coated Plates > White, 96-well

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.