Learn More
Memmert™ Plug-In Tube Extension for UF75, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand: Memmert™ UF75B29719
Beschreibung
- Accessory
- Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
- Straight
Spezifikation
UF75 Oven | |
Plug-in Tube Extension | |
O.D. 60.3 mm, I.D. 57 mm |
Produktvorschläge
Customers who viewed this item also viewed.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.