Learn More
Corning™ ProCulture™ Fittings
Dual angled sidearm fittings can be used for aseptically transferring medium into or out of angled sidearm spinner flasks or for sparging the medium with gases.
281.36 € - 649.90 €
Specifications
Material | Stainless Steel |
---|---|
No. per Case | Each |
Autoclavable | Autoclavable |
Sterility | Non-sterile |
Product Code | Brand | Product Type | Description | Diameter (Metric) Tubing | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Description | Diameter (Metric) Tubing | For Use With (Equipment) | Price | Quantity & Availability | |||
10676273
|
Corning™
4519-150 |
Dual Sidearm | 2 Tubes, Angled | 3.17 mm | 1 L Sidearm Spinner Flask |
520.67 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10184112
|
Corning™
4519-174 |
Dual Sidearm | 2 Tubes, Angled | 6.35/3.17 mm | 500 L Sidearm Spinner Flask |
576.05 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10032212
|
Corning™
4519-163 |
Combination Sidearm | 2 Tubes, Angled | 6.35 mm | 3 L Sidearm Spinner Flask |
527.51 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10377142
|
Corning™
4519-151 |
Dual Sidearm | 2 Tubes, Angled | 3.17 mm | 3 L Sidearm Spinner Flask |
527.51 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10072102
|
Corning™
4519-121 |
Dual Sidearm | 2 Tubes, Angled | 6.35/3.17 mm | 8 L Sidearm Spinner Flask |
583.18 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10718764
|
Corning™
4519-152 |
Dual Sidearm | 2 Tubes, Angled | 3.17 mm | 6 L Sidearm Spinner Flask |
541.35 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10032142
|
Corning™
4519-153 |
Dual Sidearm | 2 Tubes, Angled | 3.17 mm | 8 L Sidearm Spinner Flask |
556.67 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10511853
|
Corning™
4519-154 |
Dual Sidearm | 2 Tubes, Angled | 6.35 mm | 1 L Sidearm Spinner Flask |
520.67 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10052152
|
Corning™
4519-155 |
Dual Sidearm | 2 Tubes, Angled | 6.35 mm | 3 L Sidearm Spinner Flask |
527.51 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10307292
|
Corning™
4519-157 |
Dual Sidearm | 2 Tubes, Angled | 6.35 mm | 8 L Sidearm Spinner Flask |
556.67 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10521853
|
Corning™
4519-156 |
Dual Sidearm | 2 Tubes, Angled | 6.35 mm | 6 L Sidearm Spinner Flask |
541.35 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10377382
|
Corning™
4519-158 |
Sidearm | 2 Tubes, Angled | 3.17 mm | 1 L Sidearm Spinner Flask |
520.67 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10799524
|
Corning™
4519-160 |
Sidearm | 2 Tubes, Angled | 3.17 mm | 6 L Sidearm Spinner Flask |
541.35 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10511683
|
Corning™
4519-161 |
Sidearm | 2 Tubes, Angled | 3.17 mm | 8 L Sidearm Spinner Flask |
556.67 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10767424
|
Corning™
4519-168 |
Triple Sidearm | 3 Tubes, Angled | 3.17 mm | 6 L Sidearm Spinner Flask |
580.84 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10072162
|
Corning™
4519-159 |
Combination Sidearm | 2 Tubes, Angled | 3.17 mm | 3 L Sidearm Spinner Flask |
527.51 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10757424
|
Corning™
4519-167 |
Triple Sidearm | 3 Tubes, Angled | 3.17 mm | 3 L Sidearm Spinner Flask |
576.75 €
Each |
In Stock
Log in to see stock. |
|||
10770295
|
Corning™
4519-165 |
Sidearm | 2 Tubes, Angled | 6.35 mm | 8 L Sidearm Spinner Flask |
556.67 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10133732
|
Corning™
4519-162 |
Combination Sidearm | 2 Tubes, Angled | 6.35 mm | 1 L Sidearm Spinner Flask |
520.67 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10285792
|
Corning™
4519-166 |
Triple Sidearm | 3 Tubes, Angled | 3.17 mm | 1 L Sidearm Spinner Flask |
563.63 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10482802
|
Corning™
4519-169 |
Triple Sidearm | 3 Tubes, Angled | 3.17 mm | 8 L Sidearm Spinner Flask |
587.79 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10012122
|
Corning™
4519-170 |
Dual Sidearm | 2 Tubes, Angled | 6.35 mm | 15 L Sidearm Spinner Flask |
629.91 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10777614
|
Corning™
4519-171 |
Sidearm | 2 Tubes, Angled | 6.35 mm | 15 L Sidearm Spinner Flask |
629.91 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10550723
|
Corning™
4519-176 |
Media Handling Dual Sidearm | 2 Tubes, Straight | 6.35 mm | 8, 15 L Sidearm Spinner Flask |
649.90 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10443382
|
Corning™
4519-172 |
Sidearm | 1 Tube | 18 mm | Large Spinner Flasks w/ Vertical Sidearms |
281.36 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
10377812
|
Corning™
4519-173 |
Dual Sidearm | 2 Tubes, Angled | 6.35/3.17 mm | 1 L Sidearm Spinner Flask |
583.24 €
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||
Description
- Dual angled sidearm fittings can be used for aseptically transferring medium into or out of angled sidearm spinner flasks or for sparging the medium with gases.
- Fittings are comprised of a PET insert with a fluorocarbon O-Ring and a polypropylene sealing cap.
- Two 316 stainless steel tubes, which extend to the bottom of the flask, are held in place by modified polyphenylene oxide thermoplastic nuts with integrated ferrules.
- The fittings are completely autoclavable.
Spare parts for sidearm fittings, see Cat. Nos. 1395-32LTC, 1395-45LTC, 1395-LTR, and 1395LTMC
Specifications
Stainless Steel | |
Each | |
Autoclavable | |
Non-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Corning™ ProCulture™ Fittings