Learn More
Contec™ PROSAT™ Presaturated Nonwoven Wipes
BIOPHARMAGREENER_CHOICE
Use these wipes to clean laminar flow workbenches, cleanroom surface and equipment. PROSAT™ Presaturated Nonwoven Wipes feature task-specific saturation level of the solvent or disinfectant required to do the job.
Brand:  Contec™ PSPP0043
 View more versions of this product
Product Code. 15755533
Description
Description
Use these wipes to clean laminar flow workbenches, cleanroom surface and equipment. Contec™ PROSAT™ Presaturated Nonwoven Wipes feature task-specific saturation level of the solvent or disinfectant required to do the job.
  • Wipe substrate designed for the application
  • Compatible with ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
  • Ideal for wiping articles prior to pass through
  • Made of meltblown polypropylene or cellulose/polyester
  • Effective and convenient for wipe-downs of process tools and other equipment
  • Choose spunlaced 100% polyester wipers for improved cleanliness, meltblown polypropylene wipes for improved durability, or economical hydroentangled polyester/cellulose wipers
  • Solvent blend meets SEMI specifications for metals and contaminants
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pre-saturated Wipe
28 cm
23 cm
Not Autoclavable
Single-use
For interior surfaces of process tools, surface preparation, manufacturing processes
50 Wipes/Bag, 40 Bags/Cs.
70% IPA, 30% DI Water
11 in.
11 in.
Polypropylene
ISO Class 5
28 x 23 cm (11 x 9 in.)
Process tools and other equipment
1-ply
Non-sterile
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number855398-0-0-1 Certificate TypeCertificate of Compliance Date23/09/2021 Catalog NumberPSPP0043
Special Offers
Special Offers
Product Content Correction
Product Title
Contec™ PROSAT™ Presaturated Nonwoven Wipes > Dimensions (L x W): 28 x 23 cm (11 x 9 in.), Material: Polypropylene, Saturation Solution: 70% IPA, 30% DI Water, Quantity: 50 Wipes/Bag, 40 Bags/Cs.