Learn More
Pyrene-1-boronic acid, 95%, Thermo Scientific Chemicals
Description
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field. Also used as a intermediate for organic light-emitting diode(OLED).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field. Also used as a intermediate for organic light-emitting diode(OLED).
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Specifications
Specifications
Melting Point | 247°C to 251°C (literature) |
Color | Yellow |
Quantity | 1 g |
Beilstein | 7134073 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 246.07 |
Percent Purity | 95% |
Physical Form | Powder |
Chemical Name or Material | Pyrene-1-boronic acid |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
5012Q04Q | Certificate of Analysis | 25/10/2024 | H64211.06,H64211.03 |
5004Q01N | Certificate of Analysis | 23/04/2024 | H64211.06,H64211.03 |
5016F04W | Certificate of Analysis | 08/12/2021 | H64211.06,H64211.03 |
W29D003 | Certificate of Analysis | 08/12/2021 | H64211.06,H64211.03 |
Y12G015 | Certificate of Analysis | 08/12/2021 | H64211.06,H64211.03 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.