Learn More
Thermo Scientific Chemicals Quartz lid for 30ml quartz crucible, fused; ID 48mm
Brand: Thermo Scientific Chemicals 045555.KG
92.80 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Fused quartz | |
Quartz | |
30 mL | |
48mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only