Learn More
Thermo Scientific™ Agglutinating Sera, Salmonella O
Serologically identify somatic O antigens of Salmonella strains by slide agglutination test using Agglutinating Sera, Salmonella O.
Marque: Thermo Scientific™ R30958601
Description
Serologically identify somatic O antigens of Salmonella strains by slide agglutination test for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O. The sera are absorbed to remove agglutinins for other Salmonella antigens and the paracolon α agglutinin. Salmonella Somatic Agglutinating Sera (ZC series) are intended for the identification of O antigens.
- Select sera also for use as positive controls for stained Salmonella suspensions

Spécification
Positive | |
Salmonella O Antigen | |
Group B | |
2 mL |
Salmonella O Factor 27 (group B) | |
Agglutinating Sera | |
2 mL/Vial | |
Salmonella O |
Suggestions de produits
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3405541 | Certificate of Analysis | 18/02/2022 | R30958601 |
3242882 | Certificate of Analysis | 24/03/2021 | R30958601 |
2910795 | Certificate of Analysis | 24/12/2019 | R30958601 |
2833375 | Certificate of Analysis | 26/07/2019 | R30958601 |
2289772 | Certificate of Analysis | 05/07/2018 | R30958601 |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.