missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Agglutinating Sera, Salmonella O

Serologically identify somatic O antigens of Salmonella strains by slide agglutination test using Agglutinating Sera, Salmonella O.

Brand:  Thermo Scientific™ R30956801

 View more versions of this product

Product Code. 11933332

  • 163.00 € / 2mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Serologically identify somatic O antigens of Salmonella strains by slide agglutination test for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O. The sera are absorbed to remove agglutinins for other Salmonella antigens and the paracolon α agglutinin. Salmonella Somatic Agglutinating Sera (ZC series) are intended for the identification of O antigens.

  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Positive
Salmonella O Antigen
Group E
2 mL
Salmonella O Factors 3, 10, 15, 19 (group E)
Agglutinating Sera
2 mL/Vial
Salmonella O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Instructions

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3797941 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog NumberR30956801
Lot Number3702137 Certificate TypeCertificate of Analysis Date18/11/2023 Catalog NumberR30956801
Lot Number3594854 Certificate TypeCertificate of Analysis Date06/04/2023 Catalog NumberR30956801
Lot Number3544017 Certificate TypeCertificate of Analysis Date09/11/2022 Catalog NumberR30956801
Lot Number3429196 Certificate TypeCertificate of Analysis Date22/04/2022 Catalog NumberR30956801
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Agglutinating Sera, Salmonella O > Identifies Salmonella O Factors 3, 10, 15, 19 (Group E)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.