missing translation for 'onlineSavingsMsg'
Learn More

3M™ Safety Stripe Tape 5702

Ideal for marking lanes, hazardous areas, protruding equipment, and low hanging objects in factories, warehouses, and hospitals. Vivid, high visibility yellow and black diagonal stripes that lock into the vinyl to alert people about hazardous conditions.

Brand:  3M™ 7000017005

Product Code. 17247770

  • 1705.00 € / 24 rolls
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Applies instantly and is ready to use immediately
  • Vivid yellow and black stripes are locked into the vinyl for permanent high visibility
  • Stretches and conforms to curved surfaces and around corners for a tight fit
  • Resists abrasion, scuffing, moisture, and weathering for long service life
  • Used to identify and mark traffic areas and physical hazards
TRUSTED_SUSTAINABILITY
Specifications

Specifications

51 mm x 33 m
0.14 mm
Black with Yellow Stripes
32.91 m
5.1 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ Safety Stripe Tape 5702 > 51 mm x 33 m

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.