Learn More
Thermo Scientific™ DTME (dithiobismaleimidoethane)
Crosslink sulfhydryls to sulfhydryls (cysteine residues) with this long, cleavable crosslinker composed of maleimide groups and 11-atom (13.3A) disulfide spacer arm.
Brand: Thermo Scientific™ 22335
Description
Thermo Scientific™ Pierce DTME is a mid-length, maleimide crosslinker for reversible covalent conjugation between sulfhydryl groups (e.g., protein or peptide cysteines) by reduction of the disulfide bond in the center of the spacer arm.
Highlights:
- Cleavable by reducing agents
- Reactive groups: maleimide (homobifunctional)
- Reactive toward: sulfhydryl groups

Specifications
Sulfhydryl-Sulfhydryl | |
DTME | |
No | |
Upon receipt store desiccated at 4°C. | |
Ambient | |
Chemical Labeling | |
Maleimide | |
Solid | |
DMF, DMSO | |
No |
By Thiols | |
312.36 | |
13.3 Å | |
Yes | |
Pierce™ | |
Homobifunctional | |
Medium (10 to 30 Å) | |
50 mg | |
Standard | |
Crosslinker |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- DTME (dithiobismaleimidoethane)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Dithio-BisMaleimidoethane
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.