Learn More
Cytiva SFM4Transfx-293 Media
Promote transfection using lipofection and support growth of HEK 293 cultures with HyClone™ SFM4Transfx-293 Media, a serum-free, animal-derived component free media.
Brand:  Cytiva SH30860.02
 View more versions of this product
Product Code. 11536341
  • 86.90 € / 1000mL
Explore more special offers
Description
Description
  • ADCF
  • Supports high transfection efficiency
  • Metabolically designed for high cell yield and recombinant protein/vector production
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Liquid
0.2 μm Sterile-filtered
No Protein, No Phenol Red, No HEPES, No Glutamine
cGMP
SFM4Transfx-293
Poloxamer 188, Sodium Bicarbonate
1000 mL
2° to 8°C, away from light
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva SFM4Transfx-293 Media > 1L