Learn More
Thermo Scientific™ Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Maximize storage space with these shelf kits for Thermo Scientific™ Ultra-Low Temperature Freezers.
Brand:  Thermo Scientific™ RSK600SD4
 View more versions of this product
Product Code. 11840543
  • 1517.55 € / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Maximize storage space with Shelf Kits with Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers, featuring 2 in. boxes with dividers.
Features:
  • Stainless steel racks
  • Boxes
  • Cell dividers
Warranty varies by country. Please contact us for details.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100
Shelf Kit
Stainless Steel
Thermo Scientific TSX and TDE Series (28.8 cu. ft.)
Shelf Kit
Boxes (150/6 Racks)
4 Tiers
6
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers > Shelf Kit, Includes: 6 sliding drawer racks and (150) 2 in. boxes with 100 cell dividers, for use with Forma 88600; TSU600