Learn More
Ibidi μ-Slide 8 Well high μ-Pattern RGD, sqr30, pit110, hex Trial Pack
μ-Slide 8 Well high μ-Pattern RGD, sqr30, pit110, hex Trial Pack is an 8 well chambered coverslip with micropatterned surface.
Brand:  Ibidi 83803-S
Encompass_Preferred
Product Code. 17844794
  • POA / Pack of 2
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • A chambered coverslip with 8 wells and micropatterned surface for multi-cell assays with fluorescence microscopy readout
  • bottom material: #1.5 Polymer Coverslip with passivated surface (biologically inert) and RGD pattern
  • sterilized
Specifications
Specifications
Microscope Slide
8
2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Ibidi μ-Slide 8 Well high μ-Pattern RGD, sqr30, pit110, hex Trial Pack