missing translation for 'onlineSavingsMsg'
Learn More
Learn More

- Home
- Products
- Chemicals
- Salts and Inorganics
- Inorganic Salts
- Alkali Metal Salts
- Sodium sulfate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, anhydrous, 99.0-100.5% (calc. to the dried substance), Honeywell Fluka™
Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
Sodium sulfate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, anhydrous, 99.0-100.5% (calc. to the dried substance), Honeywell Fluka™
Product Code. 15641340
Shop All Honeywell Fluka Products

1 kg
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
50 kg
6 x 1 kg
6 x 2.5 kg
100 kg
This item is not returnable.
View return policy
Product Code. 15641340
Brand: Honeywell-Fluka 134641KG
This item is not returnable.
View return policy
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Chemical Identifiers
CAS | 7757-82-6 |
Molecular Formula | Na2O4S |
Molecular Weight (g/mol) | 142.04 |
MDL Number | MFCD00003504 |
InChI Key | PMZURENOXWZQFD-UHFFFAOYSA-L |
Synonym | sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1Show MoreShow Less |
PubChem CID | 24436 |
ChEBI | CHEBI:32149 |
SMILES | [Na+].[Na+].[O-]S([O-])(=O)=O |
Specifications
Physical Form | Anhydrous |
Quantity | 1 kg |
Linear Formula | Na2SO4 |
UN Number | NONH for all modes of transport |
IUPAC Name | disodium;sulfate |
Formula Weight | 142.04g/mol |
Percent Purity | 99 to 100.5% (calc. to the dried substance) |
Grade | puriss. |
Density | 2.68g/mL at 25°C |
Chemical Name or Material | Sodium Sulfate, Meets analytical specification of Ph. Eur.Bp, USP |
Product Suggestions
You may be interested in these alternatives
-
Product Code. 10192730
-
Product Code. 10746292
-
Product Code. 10032590
-
Product Code. 15651340
-
Product Code. 15632390
-
Product Code. 15642390
-
Product Code. 15653460
-
Product Code. 10206660
-
Product Code. 15622450
-
Product Code. 10647961
Customers who viewed this item also viewed. This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
Viewing 1-4 of
-
Catalog No. 10703811
382,00 € / 500g -
Catalog No. 10793611
154,00 € / 1kg -
Catalog No. 10117113
34,57 € / 100g -
Catalog No. 10112640
33,95 € / 1kg -
Catalog No. 10401841
27,10 € / 500mL -
Catalog No. 15681810
37,34 € / 25g
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Lot NumberO1370 | Certificate TypeCertificate of Analysis | Date02/07/2024 | Catalog Number13464-1KG |
Lot NumberM2300 | Certificate TypeCertificate of Analysis | Date06/04/2023 | Catalog Number13464-1KG |
Lot NumberM3530 | Certificate TypeCertificate of Compliance | Date05/01/2024 | Catalog Number13464-1KG |
- 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium sulfate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, anhydrous, 99.0-100.5% (calc. to the dried substance), Honeywell Fluka™ >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Spot an opportunity for improvement?Share a Content Correction