Learn More
Labcold™ Sparkfree Fridge Freezer
Stores ­flammable materials safely with no sources of ignition inside the chambers. Labcold™ Sparkfree Fridge Freezer provides all the routine storage required on a floor area no bigger than a standard fridge. For laboratory and general use where there is a concern that volatile, flammable materials or components might be stored now, or at some point in the future.
Brand:  Labcold™ RLFF13246/LK
 View more versions of this product
Product Code. 11528974
Description
Description
Combination sparkfree laboratory refrigerator and freezer provides all the routine storage required on a floor area no bigger than a standard fridge.
  • For laboratory and general use where there is a concern that volatile, flammable materials or components might be stored now, or at some point in the future.
  • Both refrigerated and frozen samples can be stored and accessed within the same small floor area.
  • Automatic defrosting in fridge
  • Drawers in Freezer are CFC and HCFC free
  • Two refrigeration systems - one for the freezer the other for the fridge
Specifications
Specifications
Spark-free Refrigerator Freezer
228 L
2°C to 10°C
4 (Refrigerator)
White Enamel
Sparkfree interior, 4 drawers in freezer, 4 shelves in fridge, and door lock with key
630 mm
595 mm
80.43 kg
134 L
-18°C to -23°C
4 (Freezer)
Eco-friendly
Molded Plastic
Refrigerator Freezer with Key
1965 mm
1.63kWh/24 hr.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Labcold™ Sparkfree Fridge Freezer > Model: RLFF13246-LK