Learn More
Description
Squalane influence on the vital pathway that transforms glucose into cholesterol may be pivotal. It regulates or controls the rate of synthesis of the enzyme HMG Co-A reductase, and may contribute to effective treatments for cancer and heart disease. It plays an important role as a terminator in the biomembrane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications
Specifications
Melting Point | -38°C |
Density | 0.81 |
Boiling Point | 210°C to 215°C (1 mmHg) |
Flash Point | 218°C (424°F) |
Refractive Index | 1.451 |
Quantity | 100 g |
Beilstein | 776019 |
Merck Index | 14,8767 |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 422.83 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10247825 | Certificate of Analysis | 05/09/2023 | A17931.22,A17931.36 |
10226424 | Certificate of Analysis | 23/11/2021 | A17931.22,A17931.36 |
10210559 | Certificate of Analysis | 23/11/2021 | A17931.22,A17931.36 |
10202219 | Certificate of Analysis | 23/11/2021 | A17931.22,A17931.36 |
10186337 | Certificate of Analysis | 23/11/2021 | A17931.22,A17931.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.