Learn More
BINDER™ Stacking Adapter
Stacking adapter to stack two devices.
Brand:  BINDER™ 9051-0048
 View more versions of this product
Product Code. 30238228
  • 495.00 € / Each
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Used to safely stack two identical devices
  • For use with KB 65, KB 130, KB PRO 130, KBF 130, or KBF PRO 130 models
Specifications
Specifications
KB 130, KB PRO 130, KBF 130, KBF PRO 130
Plastic
Stacking Adapter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BINDER™ Stacking Adapter > For use with KB 130, KB PRO 130, KBF 130, KBF PRO 130