Learn More
Contec™ Sterile NeutraKlean Concentrate

Contec Sterile NeutraKlean is a gentle and non-foaming, neutral detergent designed for the cleaning of life science cleanrooms.
Brand: Contec™ SBC015NK
Description
Provided as a unit dose concentrate, Contec Sterile NeutraKlean Concentrate is designed to be diluted in 5 L water of suitable quality, making it a cost effective product for larger areas of the cleanroom. With a pH after dilution between 6.5 and 8.0, Contec Sterile NeutraKlean is ideal for the low level cleaning required after maintenance and to remove disinfectant residues and general soil.
The product is 0.2 μm filtered and filled under Grade A air and bagged in a Grade C cleanroom and then gamma irradiated at no less than 25 kGy.
4 bottles are double bagged together, to provide easy entry into pharmaceutical cleanrooms.
Features & Benefits
- Neutral pH after dilution makes it suitable for use in all areas of cleanroom
- Filtered to 0.2 microns under Grade A airflow in a Grade C cleanroom ensures the product is free from contamination and particulates
- Low foaming detergent action makes it ideal for use on floors/walls and benches
- Sterile so ideal for high grade cleanrooms
- Low level of surfactant ensures it is low residue which can easily be removed
- Unit dose concentrate eliminates any dilution errors and ideal for use in large areas of cleanroom

Specifications
Neutral Detergent with Purified Water | |
Sterile | |
Cap | |
150 mL |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- CONTEC STERILE NEUTRAKLEAN CONCENTRATE
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : N,N DIMETHYLDECYLAMINE N OXIDE
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
210701796 | Certificate of Compliance | 11/03/2022 | SBC015NK |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.