Learn More
Sampling Systems™ SteriWare™ Cleaning Coupons
BIOPHARMA
SteriWare™ Cleaning Coupons are essential equipment for checking your cleaning processes. Cleaning coupons are available in a range of different materials and two standard sizes.
66.35 € - 127.00 €
Specifications
Packaging Type Individually Bagged
View More Specs
Products 20
Product Code Brand Material Sterility Length (Metric) Width (Metric) Price Quantity & Availability  
Product Code Brand Material Sterility Length (Metric) Width (Metric) Price Quantity & Availability  
17594636
BIOPHARMA
View Documents
Sampling Systems™
8352H-50
HDPE Non-sterile 50 mm 50 mm
66.35 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17504646
BIOPHARMA
View Documents
Sampling Systems™
8352H-50S
HDPE Sterile 50 mm 50 mm
84.80 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17534646
BIOPHARMA
View Documents
Sampling Systems™
8352N-50
Nylon Non-sterile 50 mm 50 mm
70.55 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17544646
BIOPHARMA
View Documents
Sampling Systems™
8352N-50S
Nylon Sterile 50 mm 50 mm
92.45 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17574646
BIOPHARMA
View Documents
Sampling Systems™
8352PP-50
Polypropylene Non-sterile 50 mm 50 mm
79.90 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17584646
BIOPHARMA
View Documents
Sampling Systems™
8352PP-50S
Polypropylene Sterile 50 mm 50 mm
104.00 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17514656
BIOPHARMA
View Documents
Sampling Systems™
8352PS-50
Polystyrene Non-sterile 50 mm 50 mm
82.90 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17524656
BIOPHARMA
View Documents
Sampling Systems™
8352PS-50S
Polystyrene Sterile 50 mm 50 mm
104.00 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17554656
BIOPHARMA
View Documents
Sampling Systems™
8352PVC-50
PVC Non-sterile 50 mm 50 mm
79.30 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17564656
BIOPHARMA
View Documents
Sampling Systems™
8352PVC-50S
PVC Sterile 50 mm 50 mm
101.00 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17574636
BIOPHARMA
View Documents
Sampling Systems™
8352H-100
HDPE Non-sterile 100 mm 100 mm
72.05 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17584636
BIOPHARMA
View Documents
Sampling Systems™
8352H-100S
HDPE Sterile 100 mm 100 mm
93.80 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17514646
BIOPHARMA
View Documents
Sampling Systems™
8352N-100
Nylon Non-sterile 100 mm 100 mm
91.35 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17524646
BIOPHARMA
View Documents
Sampling Systems™
8352N-100S
Nylon Sterile 100 mm 100 mm
111.00 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17554646
BIOPHARMA
View Documents
Sampling Systems™
8352PP-100
Polypropylene Non-sterile 100 mm 100 mm
83.50 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17564646
BIOPHARMA
View Documents
Sampling Systems™
8352PP-100S
Polypropylene Sterile 100 mm 100 mm
104.00 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17594646
BIOPHARMA
View Documents
Sampling Systems™
8352PS-100
Polystyrene Non-sterile 100 mm 100 mm
94.05 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17504656
BIOPHARMA
View Documents
Sampling Systems™
8352PS-100S
Polystyrene Sterile 100 mm 100 mm
114.00 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17534656
BIOPHARMA
View Documents
Sampling Systems™
8352PVC-100
PVC Non-sterile 100 mm 100 mm
107.00 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17544656
BIOPHARMA
View Documents
Sampling Systems™
8352PVC-100S
PVC Sterile 100 mm 100 mm
127.00 €
Pack of 50
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
Description
Description
  • The SteriWare™ Cleaning coupons are the perfect way to test and validate your cleaning regime.
  • The cleanroom production improves the purity of the final product.
  • Each coupon is 100% inspected and then individually bagged within the cleanroom.
  • Sterile coupons are also available from stock.
  • The coupons are available from stock in the popular sizes, materials and finishes.
  • Supplied with Certificate of Conformity for validation purposes.
  • Popular cleaning coupons include PP, nylon, PS, PVC and HDPE.
  • There are a wide range of other coupons available.
  • Made from high quality virgin material.
  • Individually bagged within the class 10,000 cleanroom.
  • Available with batch specific certificate of conformity.
  • Sterile versions are available with an irradiation certificate.
Specifications
Specifications
Individually Bagged
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Sampling Systems™ SteriWare™ Cleaning Coupons