missing translation for 'onlineSavingsMsg'
Learn More
TC-I 2000, Tocris Bioscience™ Product Code.: 30001989

TC-I 2000, Tocris Bioscience™

Product Code. 30001989
50 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 30001989

Brand: Tocris Bioscience 4632/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TRPM8 blocker

TRPM8 channel blocker. Inhibits icilin -induced TRPM8 channel activation in rTRPM8-expressing CHO cells (IC50 = 53 nM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1159996-20-9
Molecular Formula C23H18F4N2O
Molecular Weight (g/mol) 414.404
InChI Key ADCDUDFEGFKKQH-UHFFFAOYSA-N
Synonym n-4-fluorophenyl-1-4-trifluoromethyl phenyl-3,4-dihydroisoquinoline-2 1h-carboxamide, n-4-fluorophenyl-3,4-dihydro-1-4-trifluoromethyl phenyl-2 1h-isoquinolinecarboxamide, n-4-fluorophenyl-1-4-trifluoromethyl phenyl-3,4-dihydro-1h-isoquinoline-2-carboxamideShow MoreShow Less
PubChem CID 57326210
IUPAC Name N-(4-fluorophenyl)-1-[4-(trifluoromethyl)phenyl]-3,4-dihydro-1H-isoquinoline-2-carboxamide
SMILES C1CN(C(C2=CC=CC=C21)C3=CC=C(C=C3)C(F)(F)F)C(=O)NC4=CC=C(C=C4)F

Specifications

Color White
Quantity 50 mg
Formula Weight 414.4
Percent Purity >99%
Physical Form Solid
Chemical Name or Material TC-I 2000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TC-I 2000, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.