Learn More
Thermo Scientific™ TCBS Cholera Medium
Selectively isolate pathogenic vibrios with this selective, ready-to-use medium.
Brand: Thermo Scientific™ PO0194A
Description
Selectively isolate pathogenic Vibrio species with Thermo Scientific™ Oxoid™ TCBS (Thiosulfate Citrate Bile Salts Sucrose) Cholera Medium. The medium has shown to be the most satisfactory and reproducible formula regardless of the complex composition1. Unlike Lauryl Sulphate Tellurite Agar it does not require further additives or aseptic additions of blood. It also possesses superior growth characteristics for Vibrio species, compared with tellurite media. While inhibiting non-vibrios, it promotes rapid growth of pathogenic vibrios after overnight incubation at 35°C.
Use TCBS Cholera Medium for selective isolation of pathogenic vibrios.
- Produces reproducible results.
- No need for additives or aseptic additions of blood.
- Promotes rapid growth of pathogenic vibrios.
- Ready-to-use convenience of prepared media.
The TCBS medium conforms to the formulation of Kobayashi et al., except that it contains specially processed ox bile, free from the defects noted by Nakanishi and Kobayashi2.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
Green | |
TCBS Cholera Medium | |
United Kingdom | |
19g±2.0.g | |
10/Pk. |
2°C to 10°C | |
90mm Monoplate | |
Agar | |
10 x 90mm plates, film wrapped |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
4500103 | Certificate of Analysis | 28/03/2025 | PO0194A |
4499204 | Certificate of Analysis | 24/03/2025 | PO0194A |
4497505 | Certificate of Analysis | 14/03/2025 | PO0194A |
4497210 | Certificate of Analysis | 12/03/2025 | PO0194A |
4495854 | Certificate of Analysis | 07/03/2025 | PO0194A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.