Learn More
Tetrahydrofuran, 99.8%, for HPLC, unstabilized, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

Specifications
Specifications
Absorbance | (1 cm cell vs water), 0.02 max. at 280nm, 0.04 max. at 270nm, 0.05 max. at 260nm, 0.1 max. at 250nm, 0.26 max. at 240nm, 0.5 max. at 230nm |
Acidity | 0.0002 meq/g max. |
Alkalinity | 0.0006 meq/g max. |
Density | 0.8800g/mL |
Quantity | 500 mL |
Packaging | Glass Bottle |
Physical Form | Liquid |
Assay Percent Range | 99.8% |
Residue after Evaporation | 0.0002% max. |
Vapor Pressure | Vapor Pressure: 200mbar at 20°C |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2528343 | Certificate of Analysis | 25/02/2025 | 268290025,268299_10979_60KG,268290010,268291_10771_500KG,268295000 |
2413149 | Certificate of Analysis | 31/01/2025 | 268290025,268290010,268295000 |
2402240 | Certificate of Analysis | 31/01/2025 | 268290025,268290010,268295000 |
2402113 | Certificate of Analysis | 31/01/2025 | 268290025,268290010,268295000 |
2497796 | Certificate of Analysis | 31/01/2025 | 268290025,268290010,268295000 |
Safety and Handling



- Tetrahydrofuran
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.