Learn More
Thermo Scientific™ TR-FRET Coregulator Buffer G
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
Brand: Thermo Scientific™ PV4553
Description
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.

Specifications
TR-FRET Coregulator Buffer G, 25mL, Liquid, Store at 4°C, Proprietary Buffer, Water Soluble, Used For Pharma & Biopharma, Target & Lead Identification & Validation | |
25mL, 25 mL | |
TR-FRET Coregulator Buffer G may be shipped frozen or at room temperature. Store at 4°C. |
Liquid | |
LanthaScreen™ Nuclear Receptor Cofactor Interaction Assays | |
Approved for shipment at Room Temperature or on Wet or Dry Ice |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3028849A | Certificate of Analysis | 21/02/2025 | PV4553 |
2796281D | Certificate of Analysis | 21/02/2025 | PV4553 |
3028849B | Certificate of Analysis | 21/02/2025 | PV4553 |
2796281C | Certificate of Analysis | 04/04/2024 | PV4553 |
2796281B | Certificate of Analysis | 28/12/2023 | PV4553 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.