missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem 2000 C

Bright yellow color provides high visibility. Lightweight and durable fabric and a polymeric barrier coating offers good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure).

Brand:  DuPont™ D13494990

23.42 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 12379419

  • 24.15 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Hooded coverall with elastic face opening
  • Adhesive chin flap for tight fit around respirator
  • Stitched and over-taped seams
  • Souble self-adhesive zipper flap
  • Elastic cuffs, ankles and waist
  • Thumb loops prevent sleeves from riding up
  • 100% particle tight fabric
  • Excellent barrier against a broad range of concentrated inorganic chemicals and biological hazards
  • Antistatic treatment (inside)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hooded Coveralls
Tychem
Elastic
Thumb Loop
Elastic
Stitched, Taped
Chemicals
Tyvek™ C Fabric
Tychem™ C is used for splash or pressurized splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing
Small
Yellow
Elastic
Hooded
Double Zipper with Self-adhesive Flap
Category III, Type 3-B, 4-B, 5-B and 6-B
EN 14126, EN 1073-2, EN 1149-5
84 to 92 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DuPont™ Tychem 2000 C > Size: Small

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.