missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek 600 Plus White Hooded Coverall
Providing an ideal balance of protection, durability and comfort. DuPont™ Tyvek™ Taped Seam Hooded Coveralls are composed of flash spun high density polyethylene.
Brand: DuPont™ D13395299
848.23 EUR valid until 2025-12-31
Use promo code "24380" to get your promotional price.
This item is not returnable.
View return policy
Description
- Self-adhesive chin strap for tight seal of suit to face mask
- Stitched and over-taped seams for protection and strength
- Tyvek zipper and zipper flap for enhanced protection
- Elasticated face, wrists and ankles as well as glued-in elastic waist
- Elasticated thumb loops prevent sleeves from riding up
- Permeable to both air and water vapour, yet repels water-based liquids and aerosols
- Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
- Applications include maintenance and dismantling jobs in the nuclear industry, pharmaceutical manufacturing or research, biosecurity laboratories as well as medical applications and when exposed to biological hazards

Specifications
Hooded Coveralls | |
HDPE | |
Elastic (Glued-in) | |
Thumb Loop | |
Elastic | |
Stitched, Taped | |
Chemicals, Liquid | |
100 to 108 cm | |
174 to 182 cm |
Large | |
White | |
Elastic | |
Hooded | |
Zipper with Self-adhesive Flap and Chin Flap | |
Category III, Type 4-B, 5-B and 6-B | |
Category III, Type 4-B, 5-B and 6-B, EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), (EN 1149-5) | |
Maintenance and Dismantling Jobs in the Nuclear Industry, Pharmaceutical Manufacturing or in Research and Biosecurity Laboratories |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
DPT004 Tyvek classic + UK
File size: 381.9 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DuPont™ Tyvek 600 Plus White Hooded Coverall > Size: Large
Spot an opportunity for improvement?Share a Content Correction