Learn More
SI Analytics™ Ubbelohde Viscometers, ASTM
Determine absolute and relative kinematic viscosity of liquids with Newtonian flow behavior. SI Analytics™ Ubbelohde Viscometers, ASTM includes ring marks to ensure that they can be checked by means of manual measurements.
Brand:  SI Analytics™ 285400501
 View more versions of this product
Product Code. 10365051
  • 469.00 € / Each
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • In accordance with ISO 3105, ASTM D 2515, ASTM D 446
  • Calibrated for manual measurements
  • Suspended ball level for determination of absolute and relative kinematic viscosity of liquids with Newtonian flow behavior
  • Delivered with manufacturer’s certificate in accordance with DIN 55 350, Part 18
  • Provided with ring marks for automatic or manual measurements
  • Filling quantity: 15 to 20mL
  • Overall length: approx. 285mm
  • Recommended minimum flowthrough time is 200 seconds
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ubbelohde ASTM Viscometer
Yes
ISO 3105, ASTM D 2515, ASTM D 446
∼285 mm
15 to 20 mL
0.35 to 1 cSt
Capillary No. 0, Constant K = 0.001
0.24 mm
Determining the viscosity of Newtonian liquids
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SI Analytics™ Ubbelohde Viscometers, ASTM > Viscosity Range: 0.35 to 1mm²/s