missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Water purification

Aqua Purificator PG 8595 (AW) for receiving two water-filled or water-demineralizing cartridges

Brand:  Miele Professional™ 10087310

Product Code. 15460575

  • 1905.00 € / Each
Estimated Shipment: 08-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Suitable for VE / TE P 2800 and VE / TE P 2000
  • Connection of CoductivityMeter / FlowMeter possible
  • Outside dimensions: H 835 (820 without cover), B 300, T 600 mm
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lotus White
Washer-disinfectors
Galvanized Sheet, Lotus White
835 mm
26.5 kg
PG 8595 Aqua Purificator
Water Treatment
300 mm
600 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Miele Professional™ Water purification > Suitable for VE / TE P 2800 and VE / TE P 2000; color: white

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.