missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ Kimtech™ Prep™ ScottPure™ Critical Task Wipes
Kimtech™ Prep™ ScottPure™ Critical Task Wipes are engineered for excellent absorbency and low-lint wiping.
Brand: Kimberly-Clark™ 61930
200.85 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Low extractables (no binders or surfactants)
- Designed to work well with solvents (acid-, base- and solvent-resistant )
- They are made of low lint 100% rayon/polyester material for cloth-like softness
- For applications including clean manufacturing areas, printing and much more
- Absorbent, low-lint polyester / rayon material has low extractables
- Cloth-like wipers are acid-, base- and solvent-resistant
- Center pull roll has a poly wrap around that serves as the dispenser to protect the roll

Specifications
Medium-duty Industrial Wiper | |
178 mm | |
178 mm | |
White | |
Roll | |
1350 Pk. |
7 in. | |
7 in. | |
Polyester/Rayon Spunlace | |
178 x 178 mm | |
Wiping tasks where precision and reliability are essential, used in aerospace, clean manufacturing areas, DVD manufacturing, and printing | |
Non-sterile |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kimberly-Clark™ Kimtech™ Prep™ ScottPure™ Critical Task Wipes > Dimensions (L x W): 178 x 178 mm
Spot an opportunity for improvement?Share a Content Correction