Learn More
Trajan Scientific and Medical™ XCHANGE™ Syringes for use with MEPS BINS
With PTFE-tipped plunger
Brand: Trajan Scientific and Medical™ 2928500
This item is not returnable.
View return policy
Description
- High quality solutions for liquid handling
- For use on CTC PAL-xt
Specifications
PTFE-tipped (Plunger) | |
6.7 mm syringe barrel | |
100 μL |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Trajan Scientific and Medical™ XCHANGE™ Syringes for use with MEPS BINS > 100μL; 6.7mm O.D. syringe barrel
Spot an opportunity for improvement?Share a Content Correction