missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EDTA (0.5 M), pH 8.0

Optimize biochemistry or molecular biology applications that require a chelator for divalent cations.

Brand:  Thermo Scientific™ R1021

17.18 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.


Product Code. 10374440

  • 25.77 € / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

0.5 M EDTA, pH 8.0 is an aqueous solution of 0.22 μm membrane-filtered ethylendiaminetetraacetic acid (EDTA), sodium salt. It's ideal for biochemistry or molecular biology applications that require a chelator for divalent cations. By forming complexes with metal ions, EDTA inhibits metal-dependent enzymatic reactions. EDTA complexes metal ions at a 1:1 molar ratio.

Nucleic acids are normally stored in TE buffer that contains 1 mM EDTA, which prevents degradation of nucleic acids by metal dependent nucleases.

Applications
  • Termination of metal-dependent enzymatic reactions
  • Protection of DNA against metal-dependent nucleases
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EDTA
EDTA
5 x 1 mL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2800905 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR1021
Lot Number2800877 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR1021
Lot Number2832943 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR1021
Lot Number2836136 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR1021
Lot Number2854125 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberR1021
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ EDTA (0.5 M), pH 8.0 > 5 x 1mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.