Learn More
Cole-Parmer™ Digital Colony Counter Optional Magnifier
Use this magnifier with CC-200 Colony Counters.
Brand:  Cole-Parmer™ 14212-61
 View more versions of this product
Product Code. 12088183
  • 164.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Use this magnifier with Stuart™ SC6 Colony Counters.
  • 1.7x and 3x magnifier for CC-200 Colony Counter.
  • Pressure sensitive counting.
  • Average count facility.
  • Bright white energy saving LED lighting.
  • With BioCote antimicrobial protection.
  • Audible count confirmation.
  • Choice of light or dark background.
  • Connectivity to printer or computer.
Specifications
Specifications
The Colony Counter Stuart SCPPLUS
1 Ea.
Lens, Magnifying
BioCote Antimicrobial protection
3x
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Digital Colony Counter Optional Magnifier > Magnification: 3X